Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077737.1 | NP_001071205.1 | 16 - 17 | 2347 | 84 | |
XM_005161204.4 | XP_005161261.1 | 19 - 20 | 2636 | 84 | ||
XM_009295332.3 | XP_009293607.1 | 19 - 20 | 2556 | 84 | ||
GenBank mRNA | BC125837.1 | - | 16 - 17 | 2347 | 84 | |
BC165661.1 | - | 16 - 17 | 2134 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |