Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089476.1 | NP_001082945.1 | 4 - 5 | 849 | 93 | |
GenBank mRNA | BC139552.1 | - | 4 - 5 | 849 | 93 | |
BC171418.1 | - | 3 - 4 | 839 | 93 | ||
BC171424.1 | - | 3 - 4 | 839 | 93 | ||
FJ969446.1 | - | 4 - 5 | 882 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |