Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131713.2 | NP_571788.2 | 5 - 6 | 848 | 92 | |
GenBank mRNA | BC078368.1 | - | 1 - 2 | 140 | 92 | |
BC095063.1 | - | 5 - 6 | 92 | |||
BC164464.1 | - | 4 - 5 | 92 | |||
CU468784.1 | - | 4 - 5 | 92 | |||
EE697401.1 | - | 4 - 5 | 531 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |