Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144763.1 | NP_658909.1 | 17 - 18 | 1629 | 78 | |
GenBank mRNA | AJ306395.1 | - | 17 - 18 | 1629 | 78 | |
BC044450.1 | - | 17 - 18 | 1615 | 78 | ||
BC083507.1 | - | 17 - 18 | 1613 | 78 | ||
BC165764.1 | - | 17 - 18 | 1570 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |