Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312897.1 | NP_001299826.1 | 4 - 5 | 703 | 63 | |
NR_132439.1 | 5 - 6 | 573 | 63 | |||
GenBank mRNA | AL917417.1 | - | 1 - 2 | 28 | 63 | |
BC047798.1 | - | 5 - 6 | 573 | 63 | ||
CK689483.1 | - | 4 - 5 | 607 | 63 | ||
HM151010.1 | - | 4 - 5 | 512 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |