Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: naprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200890.2 | NP_957184.2 | 3 - 4 | 580 | 81 | |
XM_005170215.4 | XP_005170272.1 | 3 - 4 | 590 | 81 | ||
GenBank mRNA | BC063987.1 | - | 3 - 4 | 542 | 81 | |
EH587473.1 | - | 3 - 4 | 607 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |