Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrdc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004605.1 | NP_001004605.1 | 6 - 7 | 1175 | 74 | |
GenBank mRNA | BC078434.1 | - | 6 - 7 | 1175 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |