Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hapln1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007790.2 | NP_001007791.2 | 3 - 4 | 886 | 94 | |
NM_001313730.1 | NP_001300659.1 | 3 - 4 | 862 | 94 | ||
GenBank mRNA | AB167511.1 | - | 3 - 4 | 719 | 91 | |
BC163127.1 | - | 3 - 4 | 758 | 94 | ||
KM655692.1 | - | 3 - 4 | 698 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |