Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adcyap1r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013444.2 | NP_001013462.1 | 951 | 83 | ||
NM_001142925.1 | NP_001136397.1 | 951 | 83 | |||
NM_001142926.1 | NP_001136398.1 | 951 | 83 | |||
GenBank mRNA | AY738798.2 | - | 951 | 83 | ||
AY738800.2 | - | 951 | 83 | |||
AY738801.2 | - | 951 | 83 | |||
AY738802.2 | - | 951 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Danio rerio |