Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfcu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082899.1 | NP_001076368.1 | 3 - 4 | 1349 | 129 | |
GenBank mRNA | BC163066.1 | - | 3 - 4 | 1383 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |