Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131565.1 | NP_571640.1 | 5 - 6 | 254 | 63 | |
XM_005163151.3 | XP_005163208.3 | 6 - 7 | 394 | 63 | ||
GenBank mRNA | AF180889.1 | - | 5 - 6 | 254 | 63 | |
BC053304.1 | - | 6 - 7 | 277 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |