Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174862.3 | NP_777287.2 | 6 - 7 | 1376 | 66 | |
XM_005156971.4 | XP_005157028.1 | 7 - 8 | 1709 | 66 | ||
XM_005156972.4 | XP_005157029.1 | 8 - 9 | 1883 | 66 | ||
XM_005156973.4 | XP_005157030.1 | 7 - 8 | 1396 | 66 | ||
GenBank mRNA | AF349413.3 | - | 7 - 8 | 1395 | 66 | |
AJ275911.3 | - | 7 - 8 | 1440 | 66 | ||
AJ414566.1 | - | 7 - 8 | 1396 | 66 | ||
BC086848.1 | - | 6 - 7 | 1376 | 66 | ||
BC164792.1 | - | 6 - 7 | 1351 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |