Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199638.2 | NP_955932.2 | 5 - 6 | 1034 | 99 | |
GenBank mRNA | BC049337.1 | - | 5 - 6 | 751 | 99 | |
EH519797.1 | - | 1 - 2 | 136 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |