Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212665.1 | NP_997830.1 | 8 - 9 | 1165 | 64 | |
GenBank mRNA | BC045437.1 | - | 8 - 9 | 1165 | 64 | |
BC165837.1 | - | 8 - 9 | 1085 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |