Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh2aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009585.2 | NP_001009585.2 | 1 | 1571 | 136 | |
GenBank mRNA | AB160956.1 | - | 1 | 1543 | 136 | |
AB160957.1 | - | 1 | 1543 | 136 | ||
BC162555.1 | - | 1 | 1571 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |