Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh1gc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012657.3 | NP_001012675.3 | 1 | 1242 | 125 | |
GenBank mRNA | AB160965.1 | - | 1 | 1242 | 125 | |
AB160966.1 | - | 1 | 1242 | 125 | ||
AB160967.1 | - | 1 | 1242 | 125 | ||
AB160968.1 | - | 1 | 1242 | 125 | ||
AY576956.1 | - | 1 | 1242 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |