Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004677.2 | NP_001004677.2 | 4 - 5 | 423 | 73 | |
GenBank mRNA | BC081409.1 | - | 4 - 5 | 379 | 73 | |
BC155099.1 | - | 4 - 5 | 383 | 73 | ||
BC165129.1 | - | 3 - 4 | 355 | 73 | ||
EH538052.1 | - | 4 - 5 | 460 | 73 | ||
EH560251.1 | - | 2 - 3 | 432 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |