Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005970.1 | NP_001005970.1 | 1 - 2 | 256 | 82 | |
GenBank mRNA | BC083295.1 | - | 1 - 2 | 256 | 82 | |
BC165008.1 | - | 1 - 2 | 100 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |