Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rassf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009985.2 | NP_001009985.2 | 11 - 12 | 1082 | 94 | |
GenBank mRNA | BC088388.1 | - | 10 - 11 | 1158 | 94 | |
EH434513.1 | - | 1 - 2 | 289 | 94 |
Gene Symbol: LOC110440160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021480456.1 | XP_021336131.1 | 8 - 9 | 853 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |