Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smpdl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014338.1 | NP_001014360.1 | 5 - 6 | 826 | 108 | |
XM_005170176.4 | XP_005170233.1 | 5 - 6 | 108 | |||
XM_009294917.3 | XP_009293192.1 | 5 - 6 | 108 | |||
GenBank mRNA | BC091833.1 | - | 5 - 6 | 826 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |