Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017674.1 | NP_001017674.1 | 5 - 6 | 551 | 148 | |
GenBank mRNA | BC093332.1 | - | 5 - 6 | 551 | 148 | |
BC164386.1 | - | 4 - 5 | 493 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |