Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: baiap2l1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017830.1 | NP_001017830.1 | 11 - 12 | 1405 | 96 | |
XM_009299879.3 | XP_009298154.1 | 11 - 12 | 96 | |||
GenBank mRNA | BC092786.1 | - | 11 - 12 | 1405 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |