Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025499.1 | NP_001020670.1 | 7 - 8 | 982 | 87 | |
XM_021475037.1 | XP_021330712.1 | 7 - 8 | 1010 | 87 | ||
GenBank mRNA | BC097156.1 | - | 7 - 8 | 982 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |