Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: khdrbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077290.1 | NP_001070758.1 | 8 - 9 | 1000 | 108 | |
GenBank mRNA | BC124701.1 | - | 8 - 9 | 1000 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |