Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam98b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079662.1 | NP_001073130.1 | 2 - 3 | 273 | 129 | |
XM_017351682.2 | XP_017207171.1 | 2 - 3 | 342 | 129 | ||
GenBank mRNA | BC127591.1 | - | 2 - 3 | 273 | 129 | |
BC153522.1 | - | 2 - 3 | 256 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |