Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080081.1 | NP_001073550.1 | 6 - 7 | 745 | 146 | |
XM_009297718.3 | XP_009295993.1 | 6 - 7 | 146 | |||
GenBank mRNA | BC128877.1 | - | 6 - 7 | 745 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |