Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dachd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080673.1 | NP_001074142.1 | 9 - 10 | 1769 | 98 | |
XM_005167749.4 | XP_005167806.1 | 8 - 9 | 1761 | 98 | ||
XM_005167750.4 | XP_005167807.1 | 8 - 9 | 1764 | 98 | ||
GenBank mRNA | BC129504.1 | - | 9 - 10 | 1769 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |