Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131379.2 | NP_571454.2 | 10 - 11 | 1399 | 88 | |
GenBank mRNA | AF163311.1 | - | 10 - 11 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |