Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131859.3 | NP_571934.3 | 5 - 6 | 868 | 107 | |
GenBank mRNA | BC066635.1 | - | 5 - 6 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |