Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: heyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181736.1 | NP_859425.1 | 2 - 3 | 191 | 69 | |
GenBank mRNA | AJ510222.1 | - | 2 - 3 | 191 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |