Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198808.1 | NP_942103.1 | 10 - 11 | 1058 | 72 | |
XM_021478992.1 | XP_021334667.1 | 9 - 10 | 1365 | 72 | ||
GenBank mRNA | BC054569.1 | - | 10 - 11 | 1058 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |