Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198808.1 | NP_942103.1 | 4 - 5 | 379 | 103 | |
XM_021478992.1 | XP_021334667.1 | 3 - 4 | 686 | 103 | ||
GenBank mRNA | BC054569.1 | - | 4 - 5 | 379 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |