Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p2rx4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021478206.1 | XP_021333881.1 | 2 - 3 | 751 | 92 | |
GenBank mRNA | AY292652.1 | - | 2 - 3 | 285 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |