Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199985.2 | NP_956279.1 | 1 - 2 | 362 | 116 | |
GenBank mRNA | BC049328.1 | - | 1 - 2 | 462 | 116 | |
BC065606.1 | - | 1 - 2 | 362 | 116 | ||
BC155816.1 | - | 1 - 2 | 420 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |