Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alox5ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200061.2 | NP_956355.2 | 1 - 2 | 111 | 94 | |
GenBank mRNA | BC053181.1 | - | 1 - 2 | 111 | 94 | |
BC164909.1 | - | 1 - 2 | 95 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |