Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptgesl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200280.1 | NP_956574.1 | 5 - 6 | 952 | 91 | |
XM_021478331.1 | XP_021334006.1 | 5 - 6 | 623 | 91 | ||
GenBank mRNA | BC049325.1 | - | 5 - 6 | 952 | 91 | |
BC150469.1 | - | 4 - 5 | 1033 | 91 | ||
BC171675.1 | - | 5 - 6 | 935 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |