Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckmt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200697.1 | NP_956991.1 | 6 - 7 | 910 | 90 | |
XM_005165533.4 | XP_005165590.2 | 7 - 8 | 1051 | 90 | ||
XM_009301791.3 | XP_009300066.1 | 7 - 8 | 1002 | 90 | ||
GenBank mRNA | BC059437.1 | - | 6 - 7 | 910 | 90 | |
BC153620.1 | - | 7 - 8 | 917 | 90 | ||
BC154617.1 | - | 6 - 7 | 905 | 90 | ||
BC171364.1 | - | 6 - 7 | 888 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |