Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205546.1 | NP_991109.1 | 753 | 58 | ||
GenBank mRNA | BC065657.1 | - | 753 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio |