Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212601.2 | NP_997766.1 | 4 - 5 | 595 | 121 | |
GenBank mRNA | BC045967.1 | - | 4 - 5 | 578 | 121 | |
BC067338.1 | - | 4 - 5 | 538 | 121 | ||
BC165076.1 | - | 3 - 4 | 453 | 121 | ||
EH552076.1 | - | 4 - 5 | 593 | 121 | ||
EH588730.1 | - | 4 - 5 | 582 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |