Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vtg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102671.1 | NP_001096141.1 | 9 - 10 | 1268 | 75 | |
XM_021469334.1 | XP_021325009.1 | 9 - 10 | 1274 | 75 | ||
GenBank mRNA | BC133111.1 | - | 9 - 10 | 1268 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |