Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110134.1 | NP_001103604.1 | 3 - 4 | 437 | 77 | |
GenBank mRNA | AB364290.1 | - | 3 - 4 | 437 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |