Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242479.1 | NP_001229408.1 | 3 - 4 | 1919 | 68 | |
GenBank mRNA | AY343542.1 | - | 1 - 2 | 190 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |