Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497744.2 | XP_001497794.1 | 1164 | 147 | ||
GenBank mRNA | EU029663.1 | - | 3 - 4 | 408 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |