Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151664.1 | XP_025007432.1 | 5 - 6 | 675 | 93 | |
XM_421730.6 | XP_421730.1 | 5 - 6 | 569 | 93 | ||
GenBank mRNA | BX929388.1 | - | 4 - 5 | 521 | 92 | |
BX930620.1 | - | 5 - 6 | 563 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |