Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS27L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277460.1 | NP_001264389.1 | 2 - 3 | 227 | 68 | |
GenBank mRNA | BX260540.3 | - | 2 - 3 | 227 | 68 | |
BX276660.2 | - | 1 - 2 | 183 | 68 | ||
BX934988.1 | - | 2 - 3 | 127 | 68 | ||
CR354006.1 | - | 2 - 3 | 189 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |