Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416707.6 | XP_416707.3 | 18 - 19 | 7706 | 123 | |
GenBank mRNA | CR386728.1 | - | 4 - 5 | 414 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |