Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136182.2 | NP_001129654.1 | 2 - 3 | 315 | 92 | |
GenBank mRNA | BX932365.1 | - | 2 - 3 | 301 | 92 | |
CR405801.1 | - | 2 - 3 | 520 | 92 | ||
DN853252.1 | - | 2 - 3 | 315 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |