Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STKLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945935.3 | XP_004945992.1 | 12 - 13 | 1718 | 80 | |
XM_004945936.3 | XP_004945993.1 | 12 - 13 | 1715 | 80 | ||
XM_004945937.3 | XP_004945994.1 | 11 - 12 | 1614 | 80 | ||
XM_004945939.3 | XP_004945996.1 | 12 - 13 | 1438 | 80 | ||
XM_025141408.1 | XP_024997176.1 | 12 - 13 | 1432 | 80 | ||
GenBank mRNA | CR523822.1 | - | 1 - 2 | 88 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |