Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277883.1 | NP_001264812.1 | 4 - 5 | 483 | 149 | |
NM_001277884.1 | NP_001264813.1 | 4 - 5 | 534 | 149 | ||
XM_015296879.2 | XP_015152365.1 | 4 - 5 | 496 | 149 | ||
XM_025142514.1 | XP_024998282.1 | 4 - 5 | 468 | 149 | ||
XM_025142515.1 | XP_024998283.1 | 4 - 5 | 482 | 149 | ||
GenBank mRNA | BX929622.1 | - | 4 - 5 | 534 | 149 | |
BX931895.1 | - | 4 - 5 | 483 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |