Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277723.1 | NP_001264652.1 | 5 - 6 | 685 | 69 | |
XM_015284612.2 | XP_015140098.1 | 5 - 6 | 867 | 69 | ||
XM_015284613.2 | XP_015140099.1 | 6 - 7 | 844 | 69 | ||
XM_015284614.2 | XP_015140100.1 | 5 - 6 | 770 | 69 | ||
XM_025148754.1 | XP_025004522.1 | 4 - 5 | 800 | 69 | ||
XM_025148755.1 | XP_025004523.1 | 5 - 6 | 667 | 69 | ||
XM_025148756.1 | XP_025004524.1 | 4 - 5 | 812 | 69 | ||
GenBank mRNA | BX929623.2 | - | 5 - 6 | 704 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |